Phenylthiohydantoin-methionine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

P0366-1G P0366-100MG
TCP0366-001GEA 244.9 CAD
TCP0366-001G TCP0366-100MG
Phenylthiohydantoin-methionine ≥98.0% (by HPLC, total nitrogen)
Phenylthiohydantoin-methionine

CAS Number: 4370-90-5
MDL Number: MFCD06858379
Molecular Formula: C12H14N2OS2
Molecular Weight: 266.38
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 133


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 5-[2-(Methylthio)ethyl]-3-phenyl-2-thiohydantoin, PTH-methionine

Formula: C₁₂H₁₄N₂OS₂
Melting Pt: 133 °C
MDL Number: MFCD06858379
CAS Number: 4370-90-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8736f5d8c65df230215c3d6ed55dcb657b995b32&vid=8736f5d8c65df230215c3d6ed55dcb657b995b32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR