Phenylthiohydantoinaspartic acid ≥95.0% (by titrimetric analysis)
Supplier: TCI America
P0364-1G
P0364-100MG
TCP0364-001GEA
113.76
CAD
TCP0364-001G
TCP0364-100MG
Phenylthiohydantoinaspartic acid ≥95.0% (by titrimetric analysis)
Phenylthiohydantoinaspartic acid
CAS Number: 5624-13-5
MDL Number: MFCD01076622
Molecular Formula: C11H10N2O3S
Molecular Weight: 250.27
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 3-Phenyl-2-thiohydantoin-5-acetic Acid, PTH-aspartic Acid
Formula:
C₁₁H₁₀N₂O₃S |
MDL Number:
MFCD01076622 CAS Number: 5624-13-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...