2-(2-Pyridyl)ethanesulfonic acid ≥95.0% (by titrimetric analysis)

Supplier: TCI America

P0352-25G P0352-5G
TCP0352-025GEA 231.47 CAD
TCP0352-025G 76220-194
2-(2-Pyridyl)ethanesulfonic acid ≥95.0% (by titrimetric analysis)
2-(2-Pyridyl)ethanesulfonic acid

CAS Number: 68922-18-9
MDL Number: MFCD00006365
Molecular Formula: C7H9NO3S
Molecular Weight: 187.21
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 268

Formula: C₇H₉NO₃S
MW: 187.21 g/mol
Melting Pt: 268 °C
MDL Number: MFCD00006365
CAS Number: 68922-18-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6c59eff02169d9b94d542c34b60943e438b4e3e&vid=f6c59eff02169d9b94d542c34b60943e438b4e3e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR