4-Pyridinepropanol ≥97.0% (by GC, titration analysis)

Supplier: TCI America

P0351-25G P0351-5G
TCP0351-25GEA 717.32 CAD
TCP0351-25G TCP0351-5G
4-Pyridinepropanol ≥97.0% (by GC, titration analysis)
4-Pyridinepropanol

CAS Number: 2629-72-3
MDL Number: MFCD00047457
Molecular Formula: C8H11NO
Molecular Weight: 137.18
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 289
Flash Point (°C): 113
Freezing point (°C): 32


Other Product Names / Synonyms: 4-(3-Hydroxypropyl)pyridine, 3-(4-Pyridyl)propanol

Formula: C₈H₁₁NO
Boiling Pt: 289 °C
Flash Pt: 113 °C
MDL Number: MFCD00047457
CAS Number: 2629-72-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b4e32b70f6a9826558b785c1466b465f6024d67&vid=3b4e32b70f6a9826558b785c1466b465f6024d67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR