D-Alanine benzyl ester-p-toluenesulfonate ≥98.0%

Supplier: TCI America

Synonyms: D-Alanine Benzyl Ester p-Toluenesulfonate H-D-Ala-OBzl·TosOH D-Ala-OBzl·Tos, D-Alanine benzyl ester p-toluenesulphonate salt, H-D-Ala-OBzl.TosOH, D-Alanine benzyl ester p-toluenesulfonate salt, D-Alanine Benzyl Ester p-Toluenesulphonate H-D-Ala-OBzl·TosOH D-Ala-OBzl·Tos, H-D-Ala-OBzl·TosOH, H-D-Ala-OBzl.Tos-OH

A2004-25G A2004-5G
TCA2004-25GEA 233.84 CAD
TCA2004-25G TCA2004-5G
D-Alanine benzyl ester-p-toluenesulfonate ≥98.0%
D-Alanine benzyl ester-p-toluenesulfonate

CAS Number: 41036-32-2
MDL Number: MFCD00039092
Molecular Formula: C10H13NO2
Molecular Weight: 351.42
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 115
Specific rotation [a]20/D: 5 deg (C=3, MeOH)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-D-Ala-OBzl.Tos-OH

Formula: C₁₀H₁₃NO₂
MW: 351.42 g/mol
Melting Pt: 115 °C
MDL Number: MFCD00039092
CAS Number: 41036-32-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d74200546d17c5cc0bd930404745ee6d483ff92&vid=2d74200546d17c5cc0bd930404745ee6d483ff92&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR