Phthalyl-DL-glutamic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

P0338-5G P0338-25G
TCP0338-005GEA 175.22 CAD
TCP0338-005G TCP0338-025G
Phthalyl-DL-glutamic acid ≥98.0% (by HPLC, titration analysis)
Phthalyl-DL-glutamic acid

CAS Number: 6349-98-0
MDL Number: MFCD00023059
Molecular Formula: C13H11NO6
Molecular Weight: 277.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 192


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Phthaloyl-DL-glutamic Acid

Formula: C₁₃H₁₁NO₆
Melting Pt: 192 °C
MDL Number: MFCD00023059
CAS Number: 6349-98-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a62da396d617f51a4eddeab1222ce1589747f4cd&vid=a62da396d617f51a4eddeab1222ce1589747f4cd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR