DL-α-Phenylglycine 98,0 for synthesis

Supplier: TCI America

Synonyms: Dl-A-Aminophenylacetic Acid, H-DL-Phg-OH, DL-2-Phenylglycine

P0326-25G P0326-500G
TCP0326-025GEA 37.92 CAD
TCP0326-025G TCP0326-500G
DL-α-Phenylglycine 98,0 for synthesis
DL-α-Phenylglycine

CAS Number: 2835-06-5
MDL Number: MFCD00064402
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (+/-)-alpha-Aminophenylacetic Acid, H-DL-Phg-OH

Formula: C₈H₉NO₂
MW: 151.16 g/mol
MDL Number: MFCD00064402
CAS Number: 2835-06-5
EINECS: 220-608-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47f900ca45d027974da44b4443b1ec8004353327&vid=47f900ca45d027974da44b4443b1ec8004353327&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR