Ditridecyl phthalate (mixture of branched chain isomers)

Supplier: TCI America

P0307-500G P0307-25G
TCP0307-500GEA 176.96 CAD
TCP0307-500G TCP0307-025G
Ditridecyl phthalate (mixture of branched chain isomers)
Ditridecyl phthalate (mixture of branched chain isomers)

CAS Number: 75359-31-8
MDL Number: MFCD00048451
Molecular Formula: C34H58O4
Molecular Weight: 530.83
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 242
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: Phthalic Acid Ditridecyl Ester

Formula: C₃₄H₅₈O₄
Density: 0.96 (20 °C)
Flash Pt: 242 °C
MDL Number: MFCD00048451
CAS Number: 75359-31-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8fed28dcacd5112b369962495618dcb075578f97&vid=8fed28dcacd5112b369962495618dcb075578f97&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR