Diisopropyl phthalate ≥98.0%

Supplier: TCI America

Synonyms: Diisopropyl Phthalate

P0301-500ML P0301-25ML
TCP0301-500MLEA 564.06 CAD
TCP0301-500ML TCP0301-025ML
Diisopropyl phthalate ≥98.0%
Diisopropyl phthalate

CAS Number: 605-45-8
MDL Number: MFCD00053717
Molecular Formula: C14H18O4
Molecular Weight: 250.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Freezing point (°C): 20


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Phthalic Acid Diisopropyl Ester

Formula: C₁₄H₁₈O₄
MW: 250.2903 g/mol
MDL Number: MFCD00053717
CAS Number: 605-45-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7fbbb858d23b8461edb60f4c887a4e743e54b8e4&vid=7fbbb858d23b8461edb60f4c887a4e743e54b8e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR