6-Amino-2,4-dichloro-3-ethylphenol hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A2001-5G A2001-25G
TCA2001-5GEA 52.14 CAD
TCA2001-5G TCA2001-25G
6-Amino-2,4-dichloro-3-ethylphenol hydrochloride ≥98.0% (by HPLC, titration analysis)
6-Amino-2,4-dichloro-3-ethylphenol hydrochloride

CAS Number: 101819-99-2
Molecular Formula: C8H9Cl2NO
Molecular Weight: 242.52
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal

Formula: C₈H₉Cl₂NO
MW: 242.52 g/mol
CAS Number: 101819-99-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85c54f8e82f34876c264891c4dcbd89a5d386dfa&vid=85c54f8e82f34876c264891c4dcbd89a5d386dfa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR