Diisodecyl phthalate
Supplier: TCI America
P0299-500ML
P0299-25ML
TCP0299-500MLEA
107.44
CAD
TCP0299-500ML
TCP0299-025ML
Diisodecyl phthalate
Diisodecyl phthalate
CAS Number: 26761-40-0
MDL Number: MFCD00048389
Molecular Formula: C28H46O4
Molecular Weight: 446.67
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 235
Specific Gravity (20/20): 0.97
Other Product Names / Synonyms: Phthalic Acid Diisodecyl Ester
Formula:
C₂₈H₄₆O₄ Density: 0.97 (20 °C) Flash Pt: 235 °C |
MDL Number:
MFCD00048389 CAS Number: 26761-40-0 EINECS: 247-977-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...