Dicyclohexyl phthala ≥99.0%
Supplier: TCI America
P0293-500G
P0293-25G
TCP0293-500GEA
107.44
CAD
TCP0293-500G
TCP0293-025G
Dicyclohexyl phthala ≥99.0%
Dicyclohexyl phthala
CAS Number: 84-61-7
MDL Number: MFCD00003849
Molecular Formula: C20H26O4
Molecular Weight: 330.42
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 235
Melting point (°C): 65
Flash Point (°C): 207
Product Categories: Materials Chemistry, Resin Additives, Analytical Science, Phthalates (Environmental Endocrine Disruptors)
Other Product Names / Synonyms: Phthalic Acid Dicyclohexyl Ester
Formula:
C₂₀H₂₆O₄ Boiling Pt: 235 °C Melting Pt: 65 °C Flash Pt: 207 °C |
MDL Number:
MFCD00003849 CAS Number: 84-61-7 EINECS: 201-545-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...