Phosphocholine chloride calcium salt tetrahydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
P0274-25G
P0274-250G
TCP0274-025GEA
192.76
CAD
TCP0274-025G
TCP0274-250G
Phosphocholine chloride calcium salt tetrahydrate ≥98.0% (by titrimetric analysis)
Formula:
C₅H₁₃CaClNO₄P·4H₂O |
MDL Number:
MFCD00011755 CAS Number: 4826-71-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...