Triamyl phosphate ≥98.0%

Supplier: TCI America

P0265-25ML P0265-5ML
TCP0265-025MLEA 451.88 CAD
TCP0265-025ML TCP0265-5ML
Triamyl phosphate ≥98.0%
Triamyl phosphate

CAS Number: 2528-38-3
MDL Number: MFCD00015348
Molecular Formula: C15H33O4P
Molecular Weight: 308.40
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 158
Specific Gravity (20/20): 0.96


Product Categories: Materials Chemistry, Secondary Battery Research Reagents, Resin Additives
Other Product Names / Synonyms: Phosphoric Acid Triamyl Ester, Tripentyl Phosphate

Formula: C₁₅H₃₃O₄P
Boiling Pt: 158 °C
Density: 0.96 (20 °C)
MDL Number: MFCD00015348
CAS Number: 2528-38-3

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR