1-Naphthyl phosphate disodium salt
Supplier: TCI America
P0263-1G
P0263-5G
TCP0263-001GEA
131.84
CAD
TCP0263-001G
TCP0263-005G
1-Naphthyl phosphate disodium salt
1-Naphthyl phosphate disodium salt
[Substrate for Phosphatase]
CAS Number: 2183-17-7
MDL Number: MFCD00041007
Molecular Formula: C10H9O4P
Molecular Weight: 268.12
Form: Crystal
Color: White
Storage Temperature: <0°C
Other Product Names / Synonyms: 1-Naphthylphosphoric Acid Disodium Salt, Phosphoric Acid 1-Naphthyl Ester Disodium Salt
Formula:
C₁₀H₇Na₂O₄P |
MDL Number:
MFCD00041007 CAS Number: 2183-17-7 EINECS: 218-564-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...