1-Phenyl-2-urea ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

P0247-25G P0247-100G
TCP0247-025GEA 48.24 CAD
TCP0247-025G TCP0247-100G
1-Phenyl-2-urea ≥98.0% (by HPLC, total nitrogen)

CAS Number: 64-10-8
MDL Number: MFCD00007944
Molecular Formula: C7H8N2O
Molecular Weight: 136.15
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Boiling point (°C): 238
Melting point (°C): 147
Flash Point (°C): 190
Storage Temperature: 0-10°C

Formula: C₇H₈N₂O
Boiling Pt: 238 °C
Melting Pt: 147 °C
Flash Pt: 190 °C
MDL Number: MFCD00007944
CAS Number: 64-10-8
EINECS: 200-576-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c67255f1e1a2127acb450ef982162804680662c9&vid=c67255f1e1a2127acb450ef982162804680662c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR