3'-Acetoxyacetophenone ≥98.0%

Supplier: TCI America

A1996-25G A1996-5G
TCA1996-25GEA 50.56 CAD
TCA1996-25G TCA1996-5G
3'-Acetoxyacetophenone ≥98.0%
3'-Acetoxyacetophenone

CAS Number: 2454-35-5
MDL Number: MFCD00017227
Molecular Formula: C10H10O3
Molecular Weight: 178.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 147
Melting point (°C): 46

Formula: C₁₀H₁₀O₃
Boiling Pt: 147 °C
Melting Pt: 46 °C
MDL Number: MFCD00017227
CAS Number: 2454-35-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64387ddc188dad90a552d26c9eb1bd5dd748fdc3&vid=64387ddc188dad90a552d26c9eb1bd5dd748fdc3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR