Phenyl-p-toluenesulfonate ≥99.0%
Supplier: TCI America
Synonyms:
p-Toluenesulfonic acid Phenyl Ester, p-Toluenesulphonic acid Phenyl Ester
P0239-10G
P0239-25G
TCP0239-010GEA
61.62
CAD
TCP0239-010G
TCP0239-025G
Phenyl-p-toluenesulfonate ≥99.0%
Phenyl-p-toluenesulfonate
CAS Number: 640-60-8
MDL Number: MFCD00025996
Molecular Formula: C13H12O3S
Molecular Weight: 248.30
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 96
Other Product Names / Synonyms: p-Toluenesulfonic Acid Phenyl Ester
Formula:
C₁₃H₁₂O₃S MW: 248.30 g/mol Melting Pt: 96 °C |
MDL Number:
MFCD00025996 CAS Number: 640-60-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...