Phenyl-2-pyridyl ketone oxime ≥98.0% (by titrimetric analysis)
Supplier: TCI America
P0223-1G
P0223-5G
TCP0223-001GEA
50.56
CAD
TCP0223-001G
TCP0223-005G
Phenyl-2-pyridyl ketone oxime ≥98.0% (by titrimetric analysis)
Phenyl 2-pyridyl ketone oxime
CAS Number: 1826-28-4
MDL Number: MFCD00006310
Molecular Formula: C12H10N2O
Molecular Weight: 198.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 152
Formula:
C₁₂H₁₀N₂O Melting Pt: 152 °C |
MDL Number:
MFCD00006310 CAS Number: 1826-28-4 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...