Phenyl-1-Propenyl Ketone ≥80.0%

Supplier: TCI America

Synonyms: 1-Phenyl-2-buten-1-one

P0216-10G P0216-25G
TCP0216-010GEA 522.98 CAD
TCP0216-010G TCP0216-25G
Phenyl-1-Propenyl Ketone ≥80.0%
Phenyl-1-Propenyl Ketone

CAS Number: 495-41-0
MDL Number: MFCD00059386
Molecular Formula: C10H10O
Molecular Weight: 146.19
Purity/Analysis Method: >80.0% (GC)
Form: Clear Liquid
Boiling point (°C): 122
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: 1-Phenyl-2-buten-1-one

Formula: C₁₀H₁₀O
MW: 146.19 g/mol
Boiling Pt: 122 °C
Density: 1.03 (20 °C)
MDL Number: MFCD00059386
CAS Number: 495-41-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04367f73903b91f7557e08b70cc5c7ef8aa674af&vid=04367f73903b91f7557e08b70cc5c7ef8aa674af&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR