1-Phenyl-2-propanol ≥98.0%

Supplier: TCI America

Synonyms: (2-Hydroxypropyl)benzol, (±)-1-Phenyl-2-propanol, (±)-α-Methylphenethyl alcohol, Benzylmethylcarbinol

P0215-25G P0215-5G
TCP0215-025GEA 268.44 CAD
TCP0215-025G TCP0215-5G
1-Phenyl-2-propanol ≥98.0%
1-Phenyl-2-propanol

CAS Number: 698-87-3
MDL Number: MFCD00004540
Molecular Formula: C9H12O
Molecular Weight: 136.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 213
Flash Point (°C): 85
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: Benzylmethylcarbinol

Formula: C₆H₅CH₂CH(OH)CH₃
MW: 136.19 g/mol
Boiling Pt: 213 °C
Melting Pt: 65…67 °C
Density: 0.99 (20 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00004540
CAS Number: 698-87-3
EINECS: 211-821-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c0a4755ed8282e012eea5a0355eb7860d27dc80&vid=5c0a4755ed8282e012eea5a0355eb7860d27dc80&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR