1-(Acryloyloxy)-3-(methacryloyloxy)-2-propanol ≥80.0% stabilised

Supplier: TCI America

Synonyms: 3-(Acryloyloxy)-2-hydroxypropyl Methacrylate

A1988-25G A1988-500G
TCA1988-25GEA 33.18 CAD
TCA1988-25G TCA1988-500G
1-(Acryloyloxy)-3-(methacryloyloxy)-2-propanol ≥80.0% stabilised
1-(Acryloyloxy)-3-(methacryloyloxy)-2-propanol

(stabilized with MEHQ)
CAS Number: 1709-71-3
MDL Number: MFCD00274303
Molecular Formula: C10H14O5
Molecular Weight: 214.22
Purity/Analysis Method: >80.0% (GC)
Form: Clear Liquid
Flash Point (°C): 158
Specific Gravity (20/20): 1.14


Product Categories: Materials Chemistry, Monomers, Monomers, Monomers
Other Product Names / Synonyms: 3-(Acryloyloxy)-2-hydroxypropyl Methacrylate

Formula: C₁₀H₁₄O₅
MW: 214.22 g/mol
Density: 1.14 (20 °C)
Flash Pt: 158 °C
MDL Number: MFCD00274303
CAS Number: 1709-71-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52f8683f511b209331d4dbc00dfeb4ccbf4b4f0c&vid=52f8683f511b209331d4dbc00dfeb4ccbf4b4f0c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR