N-Benzylaniline ≥98.0%
Supplier: TCI America
P0159-25G
P0159-500G
P0159-100G
TCP0159-025GEA
73.93
CAD
TCP0159-025G
TCP0159-500G
TCP0159-100G
N-Benzylaniline ≥98.0%
N-Benzylaniline
CAS Number: 103-32-2
MDL Number: MFCD00003018
Molecular Formula: C13H13N
Molecular Weight: 183.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 307
Melting point (°C): 37
Flash Point (°C): 113
Other Product Names / Synonyms: N-Benzylaniline
Formula:
C₆H₅CH₂NHC₆H₅ Boiling Pt: 307 °C Melting Pt: 37 °C Density: 1.06 g/cm³ (20 °C) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003018 CAS Number: 103-32-2 EINECS: 203-100-4 Merck Index: 12,01161 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...