Pinacyanol chloride ≥93.0% (by HPLC)

Supplier: TCI America

P0156-1G P0156-100MG
TCP0156-001GEA 144.66 CAD
TCP0156-001G TCP0156-100MG
Pinacyanol chloride ≥93.0% (by HPLC)
Pinacyanol chloride

CAS Number: 2768-90-3
MDL Number: MFCD00011974
Molecular Formula: C25H25ClN2
Molecular Weight: 388.94
Purity/Analysis Method: >93.0% (HPLC)
Form: Crystal
Melting point (°C): 270


Product Categories: Materials Chemistry, Cyanine & Squarylium Dyes (Functional Organic Dyes), Bioscience, Stains & Dyes, Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: 1,1'-Diethyl-2,2'-carbocyanine Chloride, Quinaldine Blue

Formula: C₂₅H₂₅ClN₂
Melting Pt: 270 °C
MDL Number: MFCD00011974
CAS Number: 2768-90-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=577d13fc8b1bb8c2600905c5f22bdeb58a6a2db7&vid=577d13fc8b1bb8c2600905c5f22bdeb58a6a2db7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR