(±)-Phenylsuccinic acid ≥98.0% for synthesis

Supplier: TCI America

Synonyms: (±)-Phenyl-1,4-butanedioic acid, Phenylsuccinic acid

P0155-25G P0155-500G
TCP0155-025GEA 45.82 CAD
TCP0155-025G TCP0155-500G
(±)-Phenylsuccinic acid ≥98.0% for synthesis
(±)-Phenylsuccinic acid

CAS Number: 635-51-8
MDL Number: MFCD00004256
Molecular Formula: C10H10O4
Molecular Weight: 194.19
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 168

Formula: C₁₀H₁₀O₄
Boiling Pt: 368 °C (1013 hPa)
Melting Pt: 168 °C
MDL Number: MFCD00004256
CAS Number: 635-51-8
EINECS: 211-238-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad79865bc2fa4a49b7cbf85a8d6adb5a97f4ea7b&vid=ad79865bc2fa4a49b7cbf85a8d6adb5a97f4ea7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR