4-(Phenylazo)benzoic acid ≥98.0%
Supplier: TCI America
Synonyms:
Azobenzene-4-carboxylic Acid
P0143-5G
P0143-1G
TCP0143-005GEA
205.72
CAD
TCP0143-005G
TCP0143-1G
4-(Phenylazo)benzoic acid ≥98.0%
4-(Phenylazo)benzoic acid
CAS Number: 1562-93-2
MDL Number: MFCD00020348
Molecular Formula: C13H10N2O2
Molecular Weight: 226.24
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 248
Other Product Names / Synonyms: Azobenzene-4-carboxylic Acid
Formula:
C₁₃H₁₀N₂O₂ MW: 226.24 g/mol Melting Pt: 248 °C |
MDL Number:
MFCD00020348 CAS Number: 1562-93-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...