Phenylacetamide ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: 2-Phenylacetamide

P0120-500G P0120-25G
TCP0120-500GEA 529.3 CAD
TCP0120-500G TCP0120-025G
Phenylacetamide ≥98.0% (by total nitrogen basis)
Phenylacetamide

CAS Number: 103-81-1
MDL Number: MFCD00059193
Molecular Formula: C8H9NO
Molecular Weight: 135.17
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 156


Other Product Names / Synonyms: alpha-Toluamide

Formula: C₈H₉NO
MW: 135.17 g/mol
Melting Pt: 156 °C
MDL Number: MFCD00059193
CAS Number: 103-81-1
EINECS: 203-147-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=164040ad4351aae3c277498adbc5e90eb0561a72&vid=164040ad4351aae3c277498adbc5e90eb0561a72&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR