Phenoxyacetic anhydride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Phenoxyacetic anhydride

P0111-250G P0111-25G
TCP0111-250GEA 1640.32 CAD
TCP0111-250G TCP0111-025G
Phenoxyacetic anhydride ≥98.0% (by titrimetric analysis)
Phenoxyacetic anhydride

CAS Number: 14316-61-1
MDL Number: MFCD00059681
Molecular Formula: C16H14O5
Molecular Weight: 286.28
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 73

Formula: C₁₆H₁₄O₅
MW: 286.2794 g/mol
Melting Pt: 73 °C
MDL Number: MFCD00059681
CAS Number: 14316-61-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6c46965d8a1797c37596bfdb5d34cbccd456dc1&vid=c6c46965d8a1797c37596bfdb5d34cbccd456dc1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR