Methyl phenoxyacetate ≥99.0%

Supplier: TCI America

P0110-25G P0110-500G
TCP0110-025GEA 191.18 CAD
TCP0110-025G TCP0110-500G
Methyl phenoxyacetate ≥99.0%
Methyl phenoxyacetate

CAS Number: 2065-23-8
MDL Number: MFCD00010227
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 245
Flash Point (°C): 111
Specific Gravity (20/20): 1.15


Other Product Names / Synonyms: Phenoxyacetic Acid Methyl Ester

Formula: C₉H₁₀O₃
Boiling Pt: 245 °C
Density: 1.15 (20 °C)
Flash Pt: 111 °C
MDL Number: MFCD00010227
CAS Number: 2065-23-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68ed565c3372e7d09efb43d890204284cff10b46&vid=68ed565c3372e7d09efb43d890204284cff10b46&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR