Phenolphthalein disulfate tripotassium salt hydrate ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Phenolphthalein disulphate potassium salt hydrate, Phenolphthalein disulfate potassium salt hydrate
P0097-1G
P0097-5G
TCP0097-001GEA
328.96
CAD
TCP0097-001G
TCP0097-5G
Phenolphthalein disulfate tripotassium salt hydrate ≥97.0% (by titrimetric analysis)
Phenolphthalein disulphate tripotassium salt hydrate
CAS Number: 62625-16-5
MDL Number: MFCD00070538
Molecular Formula: C20H14O10S2
Molecular Weight: 610.73
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Formula:
C₂₀H₁₃K₃O₁₁S₂·xH₂O MW: 610.75 g/mol (anhy) |
MDL Number:
MFCD00014730 CAS Number: 62625-16-5 |
Specification Test Results
Purity (Nonaqueous Titration) | min. 97.0 % (calcd.on anh.substance) |
Water | 2.5 to 12.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...