Phenolphthalin ≥98.0%

Supplier: TCI America

Synonyms: 2-(Bis(4-hydroxyphenyl)methyl)benzoic acid

P0095-25G P0095-500G
TCP0095-025GEA 162.49 CAD
TCP0095-025G TCP0095-500G
Phenolphthalin ≥98.0%
Phenolphthalin

[for redox indicator]
CAS Number: 81-90-3
MDL Number: MFCD00036135
Molecular Formula: C20H16O4
Molecular Weight: 320.34
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 235

Formula: C₂₀H₁₆O₄
Melting Pt: 235 °C
MDL Number: MFCD00036135
CAS Number: 81-90-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1fcf2d00f39bc6546c5b79baaa08573b6d7f6449&vid=b4fbea5e7b6ddd9367a29f6c824046a449d1611a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR