2-Phenylethylamine hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: Phenethylamine Hydrochloride

P0086-25G P0086-500G P0086-100G
TCP0086-025GEA 98.84 CAD
TCP0086-025G TCP0086-500G TCP0086-100G
2-Phenylethylamine hydrochloride ≥98.0%
2-Phenylethylamine hydrochloride

CAS Number: 156-28-5
MDL Number: MFCD00012893
Molecular Formula: C8H11N
Molecular Weight: 157.64
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 224


Other Product Names / Synonyms: Phenethylamine Hydrochloride

Formula: C₈H₁₁N
MW: 157.64 g/mol
Melting Pt: 224 °C
MDL Number: MFCD00012893
CAS Number: 156-28-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6da28cbe951c2eef234762b278a29ffd05e1a717&vid=6da28cbe951c2eef234762b278a29ffd05e1a717&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR