2-Amino-2'-nitrodiphenyl sulfide ≥98.0%

Supplier: TCI America

Synonyms: 2-Aminophenyl 2-Nitrophenyl Sulfide, 2-Aminophenyl 2-Nitrophenyl sulphide, 2-(2-Nitrophenylthio)aniline

A1985-25G A1985-5G
TCA1985-25GEA 150.42 CAD
TCA1985-25G TCA1985-5G
2-Amino-2'-nitrodiphenyl sulfide ≥98.0%
2-Amino-2'-nitrodiphenyl sulfide

CAS Number: 19284-81-2
MDL Number: MFCD00121816
Molecular Formula: C12H10N2O2S
Molecular Weight: 246.28
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 87


Other Product Names / Synonyms: 2-(2-Nitrophenylthio)aniline, 2-Aminophenyl 2-Nitrophenyl Sulfide

Formula: C₁₂H₁₀N₂O₂S
MW: 246.28 g/mol
Melting Pt: 87 °C
MDL Number: MFCD00121816
CAS Number: 19284-81-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c63fbb1317d1bf2fd5621f11a9609c68e49bf42b&vid=c63fbb1317d1bf2fd5621f11a9609c68e49bf42b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR