5-Methylphenazinium methyl sulfate ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
|
Danger
|
P0083-1G
P0083-5G
TCP0083-001GEA
55.3
CAD
TCP0083-001G
TCP0083-5G
5-Methylphenazinium methyl sulfate ≥98.0% (by HPLC, total nitrogen)
5-Methylphenazinium methyl sulphate
CAS Number: 299-11-6
MDL Number: MFCD00011923
Molecular Formula: C14H14N2O4S
Molecular Weight: 306.34
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 160
Product Categories: Bioscience, Stains & Dyes
Other Product Names / Synonyms: N-Methylphenazonium Methosulfate, N-Methylphenazonium Methyl Sulfate, Phenazine Methosulfate
Formula:
C₁₄H₁₄N₂O₄S Melting Pt: 160 °C Storage Temperature: Freezer |
MDL Number:
MFCD00011923 CAS Number: 299-11-6 EINECS: 206-072-1 Merck Index: 12,06185 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...