1,3,5-Pentanetricarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
P0075-5G
P0075-25G
TCP0075-005GEA
230.68
CAD
TCP0075-005G
TCP0075-025G
1,3,5-Pentanetricarboxylic acid ≥98.0% (by GC, titration analysis)
1,3,5-Pentanetricarboxylic acid
CAS Number: 6940-58-5
MDL Number: MFCD00020546
Molecular Formula: C8H12O6
Molecular Weight: 204.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 113
Formula:
C₈H₁₂O₆ Melting Pt: 113 °C |
MDL Number:
MFCD00020546 CAS Number: 6940-58-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...