Morin hydrate ≥90.0% (by HPLC)

Supplier: TCI America

P0041-1G P0041-5G P0041-25G
TCP0041-001GEA 77.42 CAD
TCP0041-001G TCP0041-5G TCP0041-025G
Morin hydrate ≥90.0% (by HPLC)
Morin hydrate

CAS Number: 654055-01-3
MDL Number: MFCD00006826
Molecular Formula: C15H10O7
Molecular Weight: 302.24
Purity/Analysis Method: >90.0% (HPLC)
Form: Crystal


Product Categories: Bioscience, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 2',3,4',5,7-Pentahydroxyflavone

Formula: C₁₅H₁₀O₇
MDL Number: MFCD00006826
CAS Number: 654055-01-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df6ed63dbaed5ee11bd2c097a9aaef491e8781ae&vid=df6ed63dbaed5ee11bd2c097a9aaef491e8781ae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR