Pentacene, purified by sublimation
Supplier: TCI America
P0030-1G
P0030-100MG
TCP0030-001GEA
470.84
CAD
TCP0030-001G
TCP0030-100MG
Pentacene, purified by sublimation
Pentacene
CAS Number: 135-48-8
MDL Number: MFCD00003710
Molecular Formula: C22H14
Molecular Weight: 278.35
Form: Crystal
Color: Purple
Formula:
C₂₂H₁₄ Melting Pt: 300 °C Density: 1.3 g/cm³ (20 °C) |
MDL Number:
MFCD00003710 CAS Number: 135-48-8 EINECS: 205-193-7 Merck Index: 12,07240 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...