Pararosaniline ≥97.0% (by titrimetric analysis)

Supplier: TCI America

P0021-25G P0021-500G
TCP0021-025GEA 173.8 CAD
TCP0021-025G TCP0021-500G
Pararosaniline ≥97.0% (by titrimetric analysis)
Pararosaniline

CAS Number: 467-62-9
MDL Number: MFCD00036222
Molecular Formula: C19H19N3O
Molecular Weight: 305.38
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 205

Formula: C₁₉H₁₉N₃O
Melting Pt: 205 °C
MDL Number: MFCD00036222
CAS Number: 467-62-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6627a4fd1a0bf9d73c46da30b723e1879bca800&vid=a69e47a42bba6d26ceb9329bdf8f4277c0a40e96&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR