Sodium-D-pantothenate ≥97.0% (by total nitrogen basis)
Supplier: TCI America
P0013-250G
P0013-25G
TCP0013-250GEA
987.5
CAD
TCP0013-250G
TCP0013-025G
Sodium-D-pantothenate ≥97.0% (by total nitrogen basis)
Sodium-D-pantothenate
CAS Number: 867-81-2
MDL Number: MFCD00002767
Molecular Formula: C9H17NO5
Molecular Weight: 241.22
Purity/Analysis Method: >97.0% (N)
Form: Crystal
Melting point (°C): 124
Specific rotation [a]20/D: 27 deg (C=5, H2O)
Storage Temperature: 0-10°C
Product Categories: Bioscience, Vitamins
Other Product Names / Synonyms: D-Pantothenic Acid Sodium Salt
Formula:
C₉H₁₇NO₅ Melting Pt: 124 °C |
MDL Number:
MFCD00002767 CAS Number: 867-81-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...