Sodium palmitate ≥97.0% (by GC, titration analysis)
Supplier: TCI America
P0007-25G
P0007-500G
TCP0007-025GEA
195.92
CAD
TCP0007-025G
TCP0007-500G
Sodium palmitate ≥97.0% (by GC, titration analysis)
Sodium palmitate
CAS Number: 408-35-5
MDL Number: MFCD00002749
Molecular Formula: C16H32O2
Molecular Weight: 278.41
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Product Categories: Materials Chemistry, Anionic Surfactants, Bioscience, Lipids
Other Product Names / Synonyms: Palmitic Acid Sodium Salt
Formula:
C₁₆H₃₂O₂ |
MDL Number:
MFCD00002749 CAS Number: 408-35-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...