N-(4-Oxocyclohexyl)phthalimide ≥98.0% (by GC, total nitrogen)
Supplier: TCI America
O0365-25G
O0365-5G
TCO0365-25GEA
243
CAD
TCO0365-25G
TCO0365-5G
N-(4-Oxocyclohexyl)phthalimide ≥98.0% (by GC, total nitrogen)
N-(4-Oxocyclohexyl)phthalimide
CAS Number: 104618-32-8
MDL Number: MFCD00158659
Molecular Formula: C14H13NO3
Molecular Weight: 243.26
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Melting point (°C): 144
Other Product Names / Synonyms: 4-Phthalimidocyclohexanone
Formula:
C₁₄H₁₃NO₃ Melting Pt: 144 °C |
MDL Number:
MFCD00158659 CAS Number: 104618-32-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...