N-n-Octyl-D-glucamine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (2R,3R,4R,5S)-6-(Octylamino)hexane-1,2,3,4,5-pentaol, 1-Deoxy-1-(n-octylamino)-D-glucitol

O0358-25G O0358-5G
TCO0358-25GEA 56.88 CAD
TCO0358-25G TCO0358-5G
N-n-Octyl-D-glucamine ≥98.0% (by HPLC, titration analysis)
N-n-Octyl-D-glucamine

CAS Number: 23323-37-7
MDL Number: MFCD00134352
Molecular Formula: C14H31NO5
Molecular Weight: 293.40
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 137
Specific rotation [a]20/D: -19 deg (C=0.5, MeOH)


Other Product Names / Synonyms: 1-Deoxy-1-(n-octylamino)-D-glucitol

Formula: C₁₄H₃₁NO₅
MW: 293.40 g/mol
Melting Pt: 137 °C
MDL Number: MFCD00134352
CAS Number: 23323-37-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efd55a66a81277af3133bb5da84c3752c12f9908&vid=efd55a66a81277af3133bb5da84c3752c12f9908&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR