Oleanolic acid hydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

O0317-5G O0317-25G
TCO0317-5GEA 122.18 CAD
TCO0317-5G TCO0317-25G
Oleanolic acid hydrate ≥98.0% (by titrimetric analysis)
Oleanolic acid hydrate

CAS Number: 508-02-1
MDL Number: MFCD00064914
Molecular Formula: C30H48O3
Molecular Weight: 456.71
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 311
Specific rotation [a]20/D: 76 deg (C=1, CHCl3)

Formula: C₃₀H₄₈O₃·xH₂O
Melting Pt: 311 °C
MDL Number: MFCD00064914
CAS Number: 508-02-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3960eb2aad0fddb53e497b71692e0ca99d322c1&vid=37658cf8effb856ab24055e6a6fda492f5e0d6c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR