2-Acetyl-1-ethylpyrrole ≥98.0%

Supplier: TCI America

Synonyms: 1-(1-Ethyl-1H-pyrrol-2-yl)ethanone

A1977-5G A1977-25G
TCA1977-5GEA 31.6 CAD
TCA1977-5G TCA1977-25G
2-Acetyl-1-ethylpyrrole ≥98.0%
2-Acetyl-1-ethylpyrrole

CAS Number: 39741-41-8
MDL Number: MFCD00191234
Molecular Formula: C8H11NO
Molecular Weight: 137.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 82
Specific Gravity (20/20): 1.01

Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 82 °C
Density: 1.01 (20 °C)
MDL Number: MFCD00191234
CAS Number: 39741-41-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2330b887af3cb45edd8717692c41b39be94b2c4&vid=b2330b887af3cb45edd8717692c41b39be94b2c4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR