Octaethylene glycol monomethyl ether ≥96.0%

Supplier: TCI America

Synonyms: 2,5,8,11,14,17,20,23-Octaoxapentacosan-25-ol, mPEG8-Alcohol

O0296-1G O0296-5G
TCO0296-1GEA 246.32 CAD
TCO0296-1G TCO0296-5G
Octaethylene glycol monomethyl ether ≥96.0%
Octaethylene glycol monomethyl ether

CAS Number: 25990-96-9
MDL Number: MFCD06797152
Molecular Formula: C17H36O9
Molecular Weight: 384.47
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 216
Specific Gravity (20/20): 1.09
Storage Temperature: 0-10°C


Product Categories: Bioscience, PEGylation Reagents
Other Product Names / Synonyms: mPEG8-Alcohol

Formula: C₁₇H₃₆O₉
MW: 384.47 g/mol
Boiling Pt: 216 °C
Density: 1.09 (20 °C)
MDL Number: MFCD06797152
CAS Number: 25990-96-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=712526e107a14550f7285f823dce473bb1435252&vid=712526e107a14550f7285f823dce473bb1435252&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR