Ethyl-4-oxazolecarboxylate ≥96.0%

Supplier: TCI America

Synonyms: Ethyl oxazole-4-carboxylate

O0289-1G O0289-5G
TCO0289-1GEA 44.56 CAD
TCO0289-1G TCO0289-5G
Ethyl-4-oxazolecarboxylate ≥96.0%
Ethyl 4-oxazolecarboxylate

CAS Number: 23012-14-8
MDL Number: MFCD04114940
Molecular Formula: C6H7NO3
Molecular Weight: 141.13
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 101
Melting point (°C): 48
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 4-Oxazolecarboxylic Acid Ethyl Ester

Formula: C₆H₇NO₃
Boiling Pt: 101 °C
Melting Pt: 48 °C
MDL Number: MFCD04114940
CAS Number: 23012-14-8

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR