4-Oxazolecarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
O0288-1G
O0288-5G
TCO0288-1GEA
171.9
CAD
TCO0288-1G
TCO0288-5G
4-Oxazolecarboxylic acid ≥98.0% (by GC, titration analysis)
4-Oxazolecarboxylic acid
CAS Number: 23012-13-7
MDL Number: MFCD06797151
Molecular Formula: C4H3NO3
Molecular Weight: 113.07
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 142
Formula:
C₄H₃NO₃ Melting Pt: 142 °C |
MDL Number:
MFCD06797151 CAS Number: 23012-13-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...