(S)-(-)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol ≥99.0%

Supplier: TCI America

O0283-1G O0283-5G
TCO0283-001GEA 176.96 CAD
TCO0283-001G TCO0283-005G
(S)-(-)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol ≥99.0%
(S)-(-)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol

CAS Number: 65355-00-2
MDL Number: MFCD02093485
Molecular Formula: C20H22O2
Molecular Weight: 294.39
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 167
Specific rotation [a]20/D: -50 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis
Other Product Names / Synonyms: (S)-(-)-2,2'-Dihydroxy-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl

Formula: C₂₀H₂₂O₂
Melting Pt: 167 °C
MDL Number: MFCD02093485
CAS Number: 65355-00-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c6a258dafaf7bfc5f661297d55e71a4ff7d91fe&vid=4c6a258dafaf7bfc5f661297d55e71a4ff7d91fe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR