(R)-(+)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol ≥99.0%

Supplier: TCI America

O0282-1G O0282-5G O0282-25G
TCO0282-001GEA 158 CAD
TCO0282-001G TCO0282-005G TCO0282-25G
(R)-(+)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol ≥99.0%

CAS Number: 65355-14-8
MDL Number: MFCD02093485
Molecular Formula: C20H22O2
Molecular Weight: 294.39
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 167
Specific rotation [a]20/D: 50 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis
Other Product Names / Synonyms: (R)-(+)-2,2'-Dihydroxy-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl

Formula: C₂₀H₂₂O₂
Melting Pt: 167 °C
MDL Number: MFCD02093485
CAS Number: 65355-14-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea0c40c21eb5070d9658f970770054e404961dcd&vid=ea0c40c21eb5070d9658f970770054e404961dcd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR