(R)-(-)-5-Oxotetrahydrofuran-2-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

O0281-1G O0281-5G
TCO0281-001GEA 56.88 CAD
TCO0281-001G TCO0281-005G
(R)-(-)-5-Oxotetrahydrofuran-2-carboxylic acid ≥98.0% (by GC, titration analysis)
(R)-(-)-5-Oxotetrahydrofuran-2-carboxylic acid

CAS Number: 53558-93-3
MDL Number: MFCD00011567
Molecular Formula: C5H6O4
Molecular Weight: 130.10
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 72
Specific rotation [a]20/D: -15 deg (C=5, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(-)-gamma-Carboxy-gamma-butyrolactone, (R)-(-)-5-Oxo-2-tetrahydrofuroic Acid

Formula: C₅H₆O₄
Melting Pt: 72 °C
MDL Number: MFCD00011567
CAS Number: 53558-93-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5a21e8676e0a264de5d7b3f9a68bcc4fd523de0&vid=f5a21e8676e0a264de5d7b3f9a68bcc4fd523de0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR